Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284351.2 | NP_001271280.1 | 7 | 1439 | ||
NM_004174.3 | NP_004165.2 | 7 | 1439 | |||
GenBank mRNA | BC011555.1 | - | 5 | 974 | ||
BC101669.1 | - | 7 | 1335 | 8069175 | ||
BC101671.1 | - | 7 | 1335 | 8069177 | ||
BC143328.1 | - | 7 | 1335 | 9051833 | ||
BP270851.1 | - | 1 | 70 | |||
S83545.1 | - | 2 | 280 | |||
U28043.1 | - | 7 | 1337 |