Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SERPINB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006919.2 | NP_008850.1 | 6 | 635 | ||
GenBank mRNA | AB046399.1 | - | 6 | 556 | ||
AF428135.1 | - | 5 | 491 | |||
AF428136.1 | - | 5 | 491 | |||
AF428137.1 | - | 5 | 491 | |||
AF428138.1 | - | 5 | 491 | |||
AF428139.1 | - | 5 | 491 | |||
AJ515706.1 | - | 5 | 521 | |||
AK222746.1 | - | 6 | 565 | |||
AK222753.1 | - | 6 | 565 | |||
AK314805.1 | - | 6 | 565 | |||
AY190327.1 | - | 3 | 168 | |||
AY245778.1 | - | 5 | 491 | |||
AY245781.1 | - | 5 | 491 | |||
BC005224.1 | - | 6 | 565 | 4101988 | ||
BC008483.1 | - | 6 | 565 | |||
BT006748.1 | - | 5 | 491 | |||
EU852041.1 | - | 5 | 491 | |||
S66896.1 | - | 6 | 552 | |||
U19556.1 | - | 5 | 520 |