Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284341.1 | NP_001271270.1 | 4 | 775 | ||
NM_007270.4 | NP_009201.2 | 3 | 616 | |||
XM_011515115.2 | XP_011513417.1 | 3 | 616 | |||
GenBank mRNA | AF089745.1 | - | 3 | 287 | ||
AK075331.1 | - | 3 | 580 | |||
AK128597.1 | - | 4 | 557 | |||
AK294266.1 | - | 2 | 400 | |||
AK300328.1 | - | 4 | 738 | |||
AK304643.1 | - | 1 | 27 | |||
BC101723.1 | - | 3 | 544 | 8069229 | ||
BC112053.1 | - | 3 | 544 | 8327521 | ||
BC127737.1 | - | 1 | 666 | |||
BC150541.1 | - | 1 | 666 | |||
BC150542.1 | - | 1 | 666 |