Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032681.3 | NP_116070.2 | 4 | 730 | ||
GenBank mRNA | BC005014.1 | - | 2 | 370 | 3635781 | |
DV080293.1 | - | 3 | 344 |
Gene Symbol: TRIM51HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038174.2 | 3 | 602 | |||
GenBank mRNA | AB231737.1 | - | 3 | 330 |