Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000512.4 | NP_000503.1 | 14 | 2345 | ||
NM_001323543.1 | NP_001310472.1 | 13 | 2225 | |||
NM_001323544.1 | NP_001310473.1 | 15 | 2519 | |||
XM_005256301.3 | XP_005256358.1 | 14 | 5237 | |||
XM_011522982.2 | XP_011521284.1 | 15 | 5283 | |||
XM_017023112.1 | XP_016878601.1 | 14 | 7173 | |||
XM_017023113.1 | XP_016878602.1 | 13 | 5095 | |||
GenBank mRNA | AK021488.1 | - | 7 | 1434 | ||
AK124834.1 | - | 1 | 2665 | |||
AK131096.1 | - | 12 | 5664 | |||
BC007825.2 | - | 7 | 1405 | |||
BC050684.1 | - | 14 | 2334 | 6091623 | ||
BC056151.1 | - | 14 | 2326 | 6026248 | ||
BX649181.1 | - | 12 | 1938 |