Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042581.1 | NP_001036046.1 | 9 | 1383 | ||
NM_001042588.1 | NP_001036053.1 | 9 | 1188 | |||
NM_005701.3 | NP_005692.1 | 9 | 1488 | |||
GenBank mRNA | AF039029.1 | - | 9 | 1133 | ||
AK054641.1 | - | 10 | 1621 | |||
AK289475.1 | - | 9 | 1184 | |||
BC004203.2 | - | 9 | 1119 | 3605328 | ||
CR456811.1 | - | 8 | 1043 |