Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPYSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006426.2 | NP_006417.2 | 2 | 270 | ||
XM_005252658.3 | XP_005252715.1 | 2 | 259 | |||
XM_011539125.2 | XP_011537427.1 | 2 | 121 | |||
XM_017015482.1 | XP_016870971.1 | 2 | 1534 | |||
XM_017015483.1 | XP_016870972.1 | 2 | 1535 | |||
GenBank mRNA | AB006713.1 | - | 2 | 264 | ||
BC029280.1 | - | 2 | 154 | |||
BC136329.1 | - | 2 | 139 | 9020316 | ||
BC136330.1 | - | 2 | 139 | 9020317 | ||
Y10976.1 | - | 2 | 147 |