Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014296.2 | NP_055111.1 | 17 | 2224 | ||
XM_011533555.2 | XP_011531857.1 | 18 | 2123 | |||
XM_011533556.2 | XP_011531858.1 | 19 | 2355 | |||
XM_017006043.1 | XP_016861532.1 | 18 | 2307 | |||
GenBank mRNA | AB028639.1 | - | 17 | 2205 | ||
AK314253.1 | - | 17 | 2186 | |||
BC056202.1 | - | 17 | 2202 | 5274889 |