Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001656.3 | NP_001647.1 | 11 | 3784 | ||
NM_033227.2 | NP_150230.1 | 13 | 2299 | |||
NM_033228.2 | NP_150231.1 | 13 | 2270 | |||
GenBank mRNA | BC022510.1 | - | 11 | 3724 | 4811707 |