Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040118.2 | NP_001035207.1 | 26, 27 | 3761 | ||
NM_001135190.1 | NP_001128662.1 | 23, 24 | 3389 | |||
NM_015242.4 | NP_056057.2 | 24, 25 | 3572 | |||
GenBank mRNA | AB018325.2 | - | 24, 25 | 3549 | ||
AF411983.1 | - | 20, 21 | 2630 | |||
AJ621557.1 | - | 24, 25 | 3572 | |||
AK160383.1 | - | 22, 23 | 4239 | |||
AY049732.1 | - | 21, 22 | 3409 | |||
AY553630.1 | - | 26, 27 | 3776 | |||
BC008315.1 | - | 8, 9 | 1042 | |||
BC056401.1 | - | 12, 13 | 1415 | |||
BC140792.1 | - | 23, 24 | 3059 | 9021660 | ||
BC144562.1 | - | 24, 25 | 3242 | 9053090 |