Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199346.2 | NP_955378.1 | 2 | 469 | ||
XM_005264292.2 | XP_005264349.1 | 3 | 538 | |||
XM_006712007.3 | XP_006712070.1 | 3 | 280 | |||
XM_011532823.2 | XP_011531125.1 | 3 | 497 | |||
GenBank mRNA | AK292601.1 | - | 2 | 469 | ||
BC029523.1 | - | 2 | 273 | 5297707 | ||
BC043236.2 | - | 1 | 929 |