Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSSCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303024.1 | NP_001289953.1 | 2 | 323 | ||
NM_006396.2 | NP_006387.1 | 3 | 230 | |||
GenBank mRNA | AB001740.1 | - | 3 | 217 | ||
BC014791.2 | - | 3 | 216 | 3857068 | ||
BP351453.1 | - | 3 | 231 | |||
DR759640.1 | - | 2 | 290 |