Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288987.1 | NP_001275916.1 | 4 | 376 | ||
NM_014581.3 | NP_055396.1 | 4 | 376 | |||
NR_110242.1 | 5 | 482 | ||||
NR_110243.1 | 3 | 242 | ||||
XM_006717086.3 | XP_006717149.1 | 4 | 444 | |||
XM_017014654.1 | XP_016870143.1 | 5 | 612 | |||
XM_017014655.1 | XP_016870144.1 | 6 | 823 | |||
GenBank mRNA | AJ251026.1 | - | 4 | 367 | ||
AJ251027.1 | - | 5 | 473 | |||
AJ251028.1 | - | 3 | 233 | |||
AY358981.1 | - | 4 | 369 | |||
BC096717.1 | - | 4 | 351 | 40003080 | ||
BC098125.1 | - | 4 | 351 | 40003075 | ||
BC098151.1 | - | 4 | 351 | 40003079 | ||
BC098340.1 | - | 4 | 351 | 40003073 | ||
BX101545.1 | - | 2 | 97 | |||
HY046406.1 | - | 4 | 376 |
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM087898.1 | - | 4 | 283 |