Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178840.3 | NP_849162.1 | 2 | 650 | ||
GenBank mRNA | AK127425.1 | - | 2 | 650 | ||
BC017946.1 | - | 2 | 650 | 4295881 |