Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 8, 9 | 1359 | ||
XM_011545630.2 | XP_011543932.1 | 8, 9 | 1359 | |||
GenBank mRNA | AB209137.1 | - | 5 | 965 | ||
AF135016.1 | - | 7, 8 | 723 | |||
AF155098.1 | - | 2, 3 | 134 | |||
AL157501.1 | - | 1 | 70 | |||
BC009032.1 | - | 1 | 316 | |||
BC011180.1 | - | 2, 3 | 136 | |||
BC063429.1 | - | 8, 9 | 1114 | 5239974 | ||
BF530463.1 | - | 1 | 327 | |||
BM715684.1 | - | 2, 3 | 99 | |||
BU845905.1 | - | 1 | 522 |