Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BMF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003940.1 | NP_001003940.1 | 5 | 702 | ||
NM_001003942.1 | NP_001003942.1 | 3 | 404 | |||
NM_001003943.2 | NP_001003943.1 | 4 | 551 | |||
NM_033503.3 | NP_277038.1 | 4 | 553 | |||
GenBank mRNA | AK024472.1 | - | 4 | 563 | ||
AY029254.1 | - | 3 | 467 | |||
AY222040.1 | - | 3 | 404 | |||
AY222041.1 | - | 2 | 306 | |||
BC060783.1 | - | 5 | 700 | |||
BC069328.1 | - | 4 | 591 | 7262161 | ||
BC069505.1 | - | 4 | 537 | 7262173 | ||
BC070043.1 | - | 5 | 700 | 30344794 | ||
BC104983.1 | - | 4 | 488 | 8143986 | ||
BC104985.1 | - | 4 | 488 | 8143988 | ||
BP280415.1 | - | 4 | 546 |