Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNDC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032731.3 | NP_116120.1 | 4 | 531 | ||
GenBank mRNA | AF086210.1 | - | 4 | 382 | ||
AJ344101.1 | - | 4 | 426 | |||
AK291963.1 | - | 4 | 415 | |||
AK311853.1 | - | 4 | 386 | |||
BC006405.2 | - | 4 | 409 | 4298953 |