Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHCHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016139.3 | NP_057223.1 | 3, 4 | 669 | ||
GenBank mRNA | AF078845.1 | - | 3, 4 | 533 | ||
BC003079.2 | - | 3, 4 | 489 | 2988156 | ||
BC015639.2 | - | 3, 4 | 473 | 4637353 | ||
BC066331.1 | - | 3, 4 | 492 | 4792941 | ||
BC071985.1 | - | 3, 4 | 489 | 6426679 | ||
BC100275.1 | - | 3, 4 | 485 | 6192342 | ||
BF347122.1 | - | 543 |