Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RITA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286215.1 | NP_001273144.1 | 2, 3 | 864 | ||
NM_032848.2 | NP_116237.1 | 3, 4 | 996 | |||
XM_005253972.2 | XP_005254029.1 | 3, 4 | 968 | |||
GenBank mRNA | AK027733.1 | - | 3, 4 | 772 | ||
AK027741.1 | - | 3, 4 | 938 | |||
AK075358.1 | - | 3, 4 | 962 | |||
AK123762.1 | - | 4, 5 | 867 | |||
AK222908.1 | - | 3, 4 | 743 | |||
AY007158.1 | - | 3, 4 | 784 | |||
BC022092.2 | - | 3, 4 | 527 | 3029785 | ||
BX366456.2 | - | 2, 3 | 654 |