Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272071.1 | NP_001259000.1 | 6 | 786 | ||
NM_003916.4 | NP_003907.3 | 5 | 777 | |||
GenBank mRNA | AB015320.1 | - | 4 | 450 | ||
AF087876.1 | - | 4 | 568 | |||
AF091077.1 | - | 4 | 455 | |||
AF251295.1 | - | 5 | 533 | |||
AK125525.1 | - | 1 | 1930 | |||
AK130075.1 | - | 5 | 579 | |||
AK312594.1 | - | 6 | 585 | |||
BC001117.2 | - | 5 | 524 | 3050001 | ||
BC071867.1 | - | 5 | 526 | 4556064 | ||
BT006738.1 | - | 4 | 450 | |||
BX537780.1 | - | 5 | 473 |