Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014972.2 | NP_001014972.1 | 2 | 406 | ||
NM_001252612.1 | NP_001239541.1 | 2 | 732 | |||
NM_001252613.1 | NP_001239542.1 | 2 | 416 | |||
NM_014497.4 | NP_055312.2 | 2 | 416 | |||
XM_011532768.2 | XP_011531070.2 | 2 | 502 | |||
XM_017003809.1 | XP_016859298.1 | 2 | 507 | |||
XM_017003810.1 | XP_016859299.1 | 2 | 417 | |||
XM_017003811.1 | XP_016859300.1 | 2 | 502 | |||
XM_017003812.1 | XP_016859301.1 | 2 | 502 | |||
GenBank mRNA | AF534078.1 | - | 1 | 96 | ||
AK291198.1 | - | 2 | 380 | |||
AK293938.1 | - | 2 | 437 | |||
BC024000.1 | - | 2 | 375 | |||
BC064530.1 | - | 1 | 140 | |||
BC083513.1 | - | 2 | 375 | 5540321 | ||
BC143728.1 | - | 2 | 369 | 9052242 | ||
BP348645.1 | - | 2 | 409 | |||
BX537425.1 | - | 2 | 380 | |||
CR749322.1 | - | 2 | 729 | |||
D83032.1 | - | 2 | 408 |