Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032214.3 | NP_115590.1 | 6 | 877 | ||
NM_175077.2 | NP_778252.1 | 6 | 877 | |||
XM_017028098.1 | XP_016883587.1 | 6 | 868 | |||
GenBank mRNA | AF290985.1 | - | 5 | 454 | ||
AF290986.1 | - | 5 | 454 | |||
AF326353.1 | - | 5 | 454 | |||
AK025645.1 | - | 6 | 514 | |||
AK291513.1 | - | 6 | 857 | |||
BC042041.1 | - | 6 | 816 | 4429896 | ||
BQ052468.1 | - | 6 | 542 |