Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020451.2 | NP_065184.2 | 13 | 3675 | ||
NM_206926.1 | NP_996809.1 | 12 | 3573 | |||
GenBank mRNA | AF166125.1 | - | 11 | 3269 | ||
AJ306399.1 | - | 13 | 3675 | |||
AK172860.1 | - | 1 | 1997 | |||
AL110205.1 | - | 1 | 1414 | |||
BC015638.2 | - | 4 | 2345 | |||
BC021028.2 | - | 8 | 2878 | |||
BC042154.2 | - | 4 | 2345 |