Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other R3HDM2 SIRNA Assays ›
Gene Symbol:
R3HDM2
Gene Name:
R3H domain containing 2
Gene Aliases:
CAG6, PR01365
Chromosome Location:
Chr.12: 57253764 - 57430999 on Build GRCh38
UniGene ID:
Hs.443673
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
215972
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: R3HDM2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014925.3
NP_055740.3
18
2170
XM_005268725.3
XP_005268782.1
20
2520
XM_011538031.1
XP_011536333.1
21
2408
XM_011538033.1
XP_011536335.1
22
2462
XM_011538035.1
XP_011536337.1
21
2366
XM_011538036.1
XP_011536338.1
21
2360
XM_011538037.1
XP_011536339.1
20
2342
XM_011538038.1
XP_011536340.1
20
2312
XM_011538039.1
XP_011536341.1
20
2306
XM_011538040.1
XP_011536342.1
21
2360
XM_011538042.2
XP_011536344.1
23
2558
XM_017018998.1
XP_016874487.1
20
2384
XM_017018999.1
XP_016874488.1
23
2599
XM_017019000.1
XP_016874489.1
21
2366
XM_017019001.1
XP_016874490.1
22
2672
XM_017019002.1
XP_016874491.1
20
2336
XM_017019003.1
XP_016874492.1
20
2312
XM_017019004.1
XP_016874493.1
20
2306
XM_017019005.1
XP_016874494.1
22
2517
XM_017019006.1
XP_016874495.1
21
2410
XM_017019007.1
XP_016874496.1
21
2445
XM_017019008.1
XP_016874497.1
19
2210
XM_017019009.1
XP_016874498.1
20
2264
XM_017019010.1
XP_016874499.1
20
2338
XM_017019011.1
XP_016874500.1
23
2561
XM_017019012.1
XP_016874501.1
19
2246
XM_017019013.1
XP_016874502.1
20
2380
XM_017019014.1
XP_016874503.1
20
2415
XM_017019015.1
XP_016874504.1
20
2557
XM_017019016.1
XP_016874505.1
21
2576
XM_017019017.1
XP_016874506.1
20
2338
XM_017019018.1
XP_016874507.1
20
2304
XM_017019019.1
XP_016874508.1
21
2400
XM_017019020.1
XP_016874509.1
18
2150
XM_017019021.1
XP_016874510.1
20
2374
XM_017019022.1
XP_016874511.1
19
2284
XM_017019023.1
XP_016874512.1
19
2295
XM_017019024.1
XP_016874513.1
19
2284
XM_017019025.1
XP_016874514.1
19
2249
XM_017019026.1
XP_016874515.1
20
2289
XM_017019027.1
XP_016874516.1
19
2420
XM_017019028.1
XP_016874517.1
18
2182
XM_017019029.1
XP_016874518.1
18
2268
XM_017019030.1
XP_016874519.1
11
1549
XM_017019031.1
XP_016874520.1
14
1833
XM_017019032.1
XP_016874521.1
10
1401
XM_017019033.1
XP_016874522.1
12
1552
GenBank mRNA
AB023219.1
-
20
2527
AK293394.1
-
10
1422
AX775761.1
-
20
2527
AX775763.1
-
20
2569
BC041857.1
-
10
1982
5270511
BC104995.1
-
8
1177
8143998
BC112226.1
-
8
1177
8327694
BC143605.1
-
9
1279
9052118
Back To Top
More Information
Gene Ontology Categories:
Process(es)
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0