Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYBBP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105538.1 | NP_001099008.1 | 3, 4 | 448 | ||
NM_014520.3 | NP_055335.2 | 3, 4 | 448 | |||
XM_011523616.2 | XP_011521918.1 | 3, 4 | 429 | |||
GenBank mRNA | AF147709.1 | - | 3, 4 | 411 | ||
AK301622.1 | - | 3, 4 | 420 | |||
AK303150.1 | - | 3, 4 | 416 | |||
AK307948.1 | - | 3, 4 | 448 | |||
AL136595.1 | - | 2, 3 | 156 | |||
BC050546.1 | - | 3, 4 | 391 | 5495483 | ||
DC346952.1 | - | 3, 4 | 448 |