Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 10 | 1903 | ||
NM_003089.5 | NP_003080.2 | 10 | 1930 | |||
XM_005259178.1 | XP_005259235.1 | 4 | 963 | |||
XM_011527240.2 | XP_011525542.2 | 5 | 1149 | |||
XM_011527241.2 | XP_011525543.1 | 4 | 1116 | |||
GenBank mRNA | AK027127.1 | - | 3 | 1456 | ||
AK056808.1 | - | 3 | 3020 | |||
AK096783.1 | - | 11 | 3093 | |||
AL117399.1 | - | 2 | 2922 | |||
AL117507.1 | - | 3 | 1962 | |||
AL832819.1 | - | 3 | 3171 | |||
BC000342.2 | - | 2 | 824 | |||
BC001315.1 | - | 10 | 1603 | 3454479 | ||
M22636.1 | - | 10 | 1594 | |||
X06811.1 | - | 1 | 338 | |||
X06815.1 | - | 11 | 1619 | |||
X06816.1 | - | 1 | 465 | |||
X06819.1 | - | 1 | 100 | |||
X07401.1 | - | 4 | 969 |