Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRNP70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301069.1 | NP_001287998.1 | 10 | 1902 | ||
NM_003089.5 | NP_003080.2 | 10 | 1929 | |||
XM_005259178.1 | XP_005259235.1 | 4 | 962 | |||
XM_011527240.2 | XP_011525542.2 | 5 | 1148 | |||
XM_011527241.2 | XP_011525543.1 | 4 | 1115 | |||
GenBank mRNA | AK027127.1 | - | 3 | 1455 | ||
AK056808.1 | - | 3 | 3019 | |||
AK096783.1 | - | 11 | 3092 | |||
AL117399.1 | - | 2 | 2921 | |||
AL117507.1 | - | 3 | 1961 | |||
AL832819.1 | - | 3 | 3170 | |||
BC000342.2 | - | 2 | 823 | |||
BC001315.1 | - | 10 | 1602 | 3454479 | ||
M22636.1 | - | 10 | 1593 | |||
X06811.1 | - | 1 | 337 | |||
X06815.1 | - | 11 | 1618 | |||
X06816.1 | - | 1 | 464 | |||
X06819.1 | - | 1 | 99 | |||
X07401.1 | - | 4 | 968 |