Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278235.1 | NP_001265164.1 | 2, 3 | 271 | ||
NM_014583.3 | NP_055398.1 | 2, 3 | 271 | |||
XM_011533649.1 | XP_011531951.1 | 3, 4 | 326 | |||
GenBank mRNA | AF169284.1 | - | 2, 3 | 242 | ||
AF216709.1 | - | 2, 3 | 160 | |||
AK022176.1 | - | 2, 3 | 253 | |||
AK300803.1 | - | 2, 3 | 255 | |||
BC000646.2 | - | 2, 3 | 241 | 3343074 | ||
CN388104.1 | - | 2, 3 | 271 |