Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206673.1 | NP_001193602.1 | 4 | 401 | ||
NM_181533.3 | NP_853511.2 | 3 | 273 | |||
NM_181814.1 | NP_861535.1 | 2 | 170 | |||
XM_006720052.1 | XP_006720115.1 | 2 | 167 | |||
XM_011536472.1 | XP_011534774.1 | 4 | 471 | |||
XM_011536474.2 | XP_011534776.1 | 4 | 922 | |||
XM_017021031.1 | XP_016876520.1 | 3 | 335 | |||
GenBank mRNA | AK128464.1 | - | 2 | 167 | ||
AY311396.1 | - | 4 | 401 | |||
AY311397.1 | - | 3 | 273 | |||
AY311398.1 | - | 2 | 170 | |||
AY311399.1 | - | 3 | 273 | |||
BC034603.1 | - | 2 | 137 | 4778051 | ||
BC107141.1 | - | 2 | 170 | 40030548 | ||
BC107142.1 | - | 2 | 168 | 40030549 |