Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 10 | 1164 | ||
NM_014606.2 | NP_055421.1 | 11 | 1419 | |||
XM_005263327.3 | XP_005263384.1 | 12 | 1546 | |||
XM_005263329.3 | XP_005263386.1 | 11 | 1419 | |||
XM_005263330.3 | XP_005263387.1 | 11 | 1419 | |||
XM_005263331.1 | XP_005263388.1 | 11 | 1419 | |||
XM_005263332.3 | XP_005263389.1 | 11 | 1419 | |||
XM_017008806.1 | XP_016864295.1 | 12 | 1546 | |||
XM_017008807.1 | XP_016864296.1 | 10 | 1142 | |||
XM_017008808.1 | XP_016864297.1 | 3 | 225 | |||
GenBank mRNA | AK289990.1 | - | 11 | 1362 | ||
AK296379.1 | - | 10 | 1140 | |||
D25215.1 | - | 11 | 1346 |