Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004334.3 | NP_001004334.3 | 11 | 4698 | ||
GenBank mRNA | AY584611.1 | - | 11 | 4698 |