Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR5T2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004746.1 | NP_001004746.1 | 1 | 136 | ||
GenBank mRNA | BC136876.1 | - | 1 | 212 | 9020867 |
Set Membership: |
G-protein coupled receptor |