Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PDE4DIP SIRNA Assays ›
Gene Symbol:
PDE4DIP
Gene Name:
phosphodiesterase 4D interacting protein
Gene Aliases:
CMYA2, MMGL
Chromosome Location:
Chr.1: 148808167 - 149051277 on Build GRCh38
UniGene ID:
Hs.584841
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21652
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PDE4DIP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002811.2
NP_001002811.2
1
1232
XM_005272981.3
XP_005273038.2
1
1015
XM_005277442.4
XP_005277499.2
1
1015
XM_005277443.3
XP_005277500.1
1
1015
XM_006711642.2
XP_006711705.1
1
1015
XM_006711643.2
XP_006711706.1
1
1015
XM_006711644.2
XP_006711707.1
1
1015
XM_006711648.2
XP_006711711.1
1
1015
XM_011510174.1
XP_011508476.1
1
1015
XM_017002875.1
XP_016858364.1
1
1015
XM_017002876.1
XP_016858365.1
1
1015
XM_017002877.1
XP_016858366.1
1
1015
XM_017002879.1
XP_016858368.1
1
1015
XM_017002881.1
XP_016858370.1
1
1015
XM_017002882.1
XP_016858371.1
1
1015
XM_017002883.1
XP_016858372.1
1
1015
XM_017002884.1
XP_016858373.1
1
1015
XM_017002885.1
XP_016858374.1
1
1015
XM_017002886.1
XP_016858375.1
1
1015
XM_017002890.1
XP_016858379.1
1
1015
XM_017002896.1
XP_016858385.1
1
1015
XM_017002897.1
XP_016858386.1
1
1015
XM_017002900.1
XP_016858389.1
1
1015
XM_017002901.1
XP_016858390.1
1
1015
GenBank mRNA
AB007946.1
-
1
1232
BC004860.2
-
1
899
BC078660.1
-
1
738
BC132717.1
-
1
835
BC152439.1
-
1
1232
HQ333476.1
-
1
645
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein complex assembly
protein binding
enzyme binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0