Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169154.1 | NP_001162625.1 | 3 | 445 | ||
NM_173494.1 | NP_775765.1 | 2 | 170 | |||
XM_011530855.2 | XP_011529157.1 | 3 | 285 | |||
GenBank mRNA | AI147427.1 | - | 2 | 401 | ||
AY211928.1 | - | 2 | 165 | |||
BC033510.1 | - | 2 | 170 | 5173807 |