Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014648.3 | NP_055463.1 | 5 | 721 | ||
XM_005247914.3 | XP_005247971.1 | 5 | 520 | |||
XM_005247915.3 | XP_005247972.1 | 5 | 490 | |||
XM_005247916.3 | XP_005247973.1 | 6 | 662 | |||
XM_005247917.3 | XP_005247974.1 | 5 | 451 | |||
XM_005247918.3 | XP_005247975.1 | 5 | 938 | |||
XM_005247919.3 | XP_005247976.1 | 5 | 690 | |||
XM_017007540.1 | XP_016863029.1 | 5 | 567 | |||
GenBank mRNA | AB014575.1 | - | 5 | 428 | ||
AK023138.1 | - | 5 | 489 | |||
AK023389.1 | - | 5 | 530 | |||
AY227651.1 | - | 5 | 467 | |||
AY227654.1 | - | 5 | 506 | |||
BC039018.1 | - | 5 | 482 | |||
BC056674.1 | - | 5 | 493 | |||
BC063882.1 | - | 5 | 474 | 30387514 |