Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320865.1 | NP_001307794.1 | 9 | 964 | ||
NM_198559.1 | NP_940961.1 | 9 | 931 | |||
XM_005246539.4 | XP_005246596.1 | 8 | 928 | |||
XM_005246541.4 | XP_005246598.1 | 7 | 727 | |||
XM_011511148.2 | XP_011509450.1 | 8 | 895 | |||
XM_011511149.2 | XP_011509451.1 | 7 | 726 | |||
XM_011511150.2 | XP_011509452.1 | 7 | 692 | |||
XM_017004053.1 | XP_016859542.1 | 7 | 1127 | |||
GenBank mRNA | AK098178.1 | - | 1 | 1991 | ||
BC052750.1 | - | 9 | 931 | 5742766 |