Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014663.2 | NP_055478.2 | 5 | 660 | ||
XM_005271354.3 | XP_005271411.1 | 5 | 697 | |||
XM_005271355.3 | XP_005271412.1 | 5 | 615 | |||
XM_005271356.3 | XP_005271413.1 | 6 | 736 | |||
XM_017002912.1 | XP_016858401.1 | 6 | 805 | |||
GenBank mRNA | AB014577.1 | - | 5 | 619 | ||
AK300039.1 | - | 5 | 652 | |||
BC002558.2 | - | 5 | 628 | 3138875 |