Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014972.2 | NP_001014972.1 | 22 | 4033 | ||
NM_001252612.1 | NP_001239541.1 | 22 | 4359 | |||
NM_001252613.1 | NP_001239542.1 | 22 | 4043 | |||
NM_014497.4 | NP_055312.2 | 22 | 4043 | |||
XM_005264263.1 | XP_005264320.1 | 21 | 2507 | |||
XM_006711989.1 | XP_006712052.1 | 21 | 2519 | |||
XM_011532768.2 | XP_011531070.2 | 21 | 4003 | |||
XM_017003809.1 | XP_016859298.1 | 22 | 4134 | |||
XM_017003810.1 | XP_016859299.1 | 21 | 3918 | |||
GenBank mRNA | AF273049.1 | - | 12 | 1476 | ||
AK128706.1 | - | 3 | 2000 | |||
AK291198.1 | - | 22 | 4007 | |||
AK303723.1 | - | 20 | 2503 | |||
BC083513.1 | - | 22 | 4002 | 5540321 | ||
BC143728.1 | - | 22 | 3996 | 9052242 | ||
CR749322.1 | - | 22 | 4356 | |||
D83032.1 | - | 22 | 4035 |