Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 9 | 1174 | ||
XM_011527491.2 | XP_011525793.1 | 9 | 717 | |||
XM_017027472.1 | XP_016882961.1 | 8 | 681 | |||
GenBank mRNA | AK098197.1 | - | 3 | 836 | ||
AY444561.1 | - | 8 | 649 | |||
AY551341.1 | - | 9 | 732 | |||
BC009072.1 | - | 9 | 706 | |||
BC010736.1 | - | 9 | 727 | |||
BC050082.1 | - | 9 | 750 | |||
BC121146.1 | - | 8 | 1043 | 40121256 | ||
BC121147.1 | - | 9 | 1102 | 40121258 | ||
BM463473.1 | - | 1 | 40 |