Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 1 | 151 | ||
GenBank mRNA | BC121146.1 | - | 1 | 79 | 40121256 | |
BC121147.1 | - | 1 | 79 | 40121258 | ||
BE386959.1 | - | 1 | 153 | |||
BI856163.1 | - | 1 | 281 |