Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CFHR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166624.1 | NP_001160096.1 | 5 | 1170 | ||
NM_021023.5 | NP_066303.2 | 6 | 1353 | |||
GenBank mRNA | BC058009.1 | - | 6 | 1311 | 4716505 |
Gene Symbol: DDX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL713763.1 | - | 11 | 1806 |