Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021244.4 | NP_067067.1 | 7 | 1577 | ||
XM_005248755.4 | XP_005248812.1 | 6 | 1622 | |||
XM_011536022.2 | XP_011534324.1 | 7 | 1349 | |||
GenBank mRNA | AF272036.1 | - | 7 | 1360 | ||
AK126164.1 | - | 1 | 2653 | |||
AK289799.1 | - | 7 | 1502 | |||
AL137502.1 | - | 6 | 1075 | |||
BC003088.1 | - | 7 | 1239 | 3536088 |