Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIB2 SIRNA Assays ›
Gene Symbol:
MIB2
Gene Name:
mindbomb E3 ubiquitin protein ligase 2
Gene Aliases:
ZZANK1, ZZZ5
Chromosome Location:
Chr.1: 1615188 - 1630610 on Build GRCh38
UniGene ID:
Hs.135805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
216955
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001170686.1
NP_001164157.1
19
2987
NM_001170687.1
NP_001164158.1
19
2957
NM_001170688.1
NP_001164159.1
18
2804
NM_001170689.1
NP_001164160.1
17
2597
NM_080875.2
NP_543151.2
19
2999
NR_033183.1
19
2782
XM_006710372.1
XP_006710435.1
18
2863
XM_011540731.2
XP_011539033.1
20
3835
XM_011540736.2
XP_011539038.1
20
3736
XM_011540737.2
XP_011539039.1
19
3673
XM_011540741.2
XP_011539043.1
18
3568
XM_011540742.2
XP_011539044.1
18
3565
XM_017000349.1
XP_016855838.1
19
3907
XM_017000350.1
XP_016855839.1
19
3895
XM_017000351.1
XP_016855840.1
19
3889
XM_017000352.1
XP_016855841.1
19
3865
XM_017000353.1
XP_016855842.1
18
3844
XM_017000354.1
XP_016855843.1
19
3808
XM_017000355.1
XP_016855844.1
17
3739
XM_017000356.1
XP_016855845.1
17
3736
XM_017000357.1
XP_016855846.1
18
3712
XM_017000358.1
XP_016855847.1
19
3205
XM_017000359.1
XP_016855848.1
19
3193
XM_017000360.1
XP_016855849.1
19
3152
XM_017000361.1
XP_016855850.1
18
3043
XM_017000362.1
XP_016855851.1
19
3245
XM_017000363.1
XP_016855852.1
19
3208
XM_017000364.1
XP_016855853.1
17
2848
XM_017000365.1
XP_016855854.1
19
2935
XM_017000366.1
XP_016855855.1
18
2872
GenBank mRNA
AB074480.1
-
19
2910
AB076691.1
-
19
2868
AB076692.1
-
18
2715
AB076693.1
-
19
2898
AB097000.1
-
19
2833
AK000655.1
-
2
252
AK091610.1
-
13
1803
AK095914.1
-
13
1805
AK096295.1
-
14
1910
AK097106.1
-
15
2229
AK098785.1
-
20
2785
AK122751.1
-
19
2701
AK122863.1
-
18
2993
AK128167.1
-
17
2557
AK128553.1
-
18
3913
AK293642.1
-
18
2979
AK302757.1
-
19
2951
AL834527.1
-
14
2031
BC016490.1
-
19
2836
BC037542.1
-
19
2879
5168033
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0