Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300861.1 | NP_001287790.1 | 2 | 339 | ||
NM_020470.2 | NP_065203.2 | 2 | 339 | |||
XM_005273720.3 | XP_005273777.1 | 2 | 315 | |||
XM_017017139.1 | XP_016872628.1 | 2 | 315 | |||
XM_017017140.1 | XP_016872629.1 | 2 | 315 | |||
XM_017017141.1 | XP_016872630.1 | 2 | 315 | |||
GenBank mRNA | AF004876.1 | - | 2 | 272 | ||
AK226003.1 | - | 2 | 317 | |||
AK308988.1 | - | 2 | 309 | |||
AK310521.1 | - | 2 | 218 | |||
BC001299.1 | - | 2 | 290 | 3451489 | ||
BG397210.1 | - | 2 | 246 | |||
CR456710.1 | - | 2 | 157 | |||
DB085949.1 | - | 2 | 339 | |||
S83365.1 | - | 1 | 130 |