Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM192A SIRNA Assays ›
Gene Symbol:
FAM192A
Gene Name:
family with sequence similarity 192 member A
Gene Aliases:
C16orf94, CDA018, CDA10, NIP30
Chromosome Location:
Chr.16: 57152466 - 57186242 on Build GRCh38
UniGene ID:
Hs.396740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
217067
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM192A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024946.2
NP_079222.1
7
1079
XM_005256156.4
XP_005256213.1
7
1047
XM_005256158.3
XP_005256215.1
7
2134
XM_005256160.2
XP_005256217.1
7
1051
XM_005256163.2
XP_005256220.1
8
1188
XM_005256164.3
XP_005256221.1
8
1133
XM_005256165.2
XP_005256222.1
7
1108
XM_006721275.3
XP_006721338.2
7
1817
XM_011523343.2
XP_011521645.1
7
925
XM_017023681.1
XP_016879170.1
7
1143
XM_017023682.1
XP_016879171.1
8
2252
XM_017023683.1
XP_016879172.1
8
2203
XM_017023684.1
XP_016879173.1
8
1418
XM_017023685.1
XP_016879174.1
8
2167
XM_017023686.1
XP_016879175.1
9
2237
XM_017023687.1
XP_016879176.1
9
2212
XM_017023688.1
XP_016879177.1
9
2155
XM_017023689.1
XP_016879178.1
9
1536
XM_017023690.1
XP_016879179.1
9
1487
XM_017023691.1
XP_016879180.1
9
1540
XM_017023692.1
XP_016879181.1
8
1324
XM_017023693.1
XP_016879182.1
8
1227
XM_017023694.1
XP_016879183.1
9
1330
XM_017023695.1
XP_016879184.1
9
1442
XM_017023696.1
XP_016879185.1
8
1825
XM_017023697.1
XP_016879186.1
8
1210
XM_017023698.1
XP_016879187.1
8
1933
XM_017023699.1
XP_016879188.1
8
1461
XM_017023700.1
XP_016879189.1
8
930
XM_017023701.1
XP_016879190.1
8
1396
XM_017023702.1
XP_016879191.1
8
1116
XM_017023703.1
XP_016879192.1
7
856
XM_017023704.1
XP_016879193.1
7
854
GenBank mRNA
AF212249.1
-
7
1032
AF271785.1
-
7
1032
AF356585.1
-
6
830
AK023291.1
-
7
979
AK025452.1
-
7
1054
AL834421.1
-
7
891
BC020536.1
-
7
1047
3849620
BC063409.1
-
7
897
BC071952.1
-
6
848
6305656
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-wl7lb:80/10.239.167.53:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0