Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022742.4 | NP_073579.4 | 13 | 2856 | ||
XM_005250534.3 | XP_005250591.1 | 14 | 2640 | |||
XM_005250538.1 | XP_005250595.1 | 14 | 2640 | |||
XM_011516483.1 | XP_011514785.1 | 15 | 2712 | |||
XM_011516484.1 | XP_011514786.1 | 15 | 2712 | |||
XM_011516485.1 | XP_011514787.1 | 15 | 2709 | |||
XM_011516486.1 | XP_011514788.1 | 15 | 2676 | |||
XM_011516487.2 | XP_011514789.1 | 15 | 2712 | |||
XM_011516488.2 | XP_011514790.1 | 15 | 2712 | |||
XM_011516489.1 | XP_011514791.1 | 15 | 2712 | |||
XM_017012532.1 | XP_016868021.1 | 14 | 2604 | |||
XM_017012533.1 | XP_016868022.1 | 14 | 3106 | |||
XM_017012534.1 | XP_016868023.1 | 14 | 2604 | |||
GenBank mRNA | AB058696.1 | - | 13 | 2807 | ||
AK023500.1 | - | 6 | 1213 | |||
AL133027.1 | - | 10 | 2070 | |||
BC070114.1 | - | 10 | 2593 | |||
BC150331.1 | - | 13 | 2818 | 8860498 | ||
BC152408.1 | - | 13 | 2807 | 8862511 |