Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145862.1 | NP_001139334.1 | 17 | 2279 | ||
NM_181873.3 | NP_870988.2 | 16 | 2021 | |||
XM_006711137.1 | XP_006711200.1 | 16 | 1998 | |||
XM_011509099.2 | XP_011507401.1 | 14 | 3122 | |||
GenBank mRNA | AK097000.1 | - | 16 | 1998 | ||
AK298775.1 | - | 17 | 2248 | |||
BC032655.1 | - | 10 | 1353 | |||
BC064372.1 | - | 15 | 2240 | |||
BC094756.1 | - | 10 | 4303 | |||
BC100818.2 | - | 16 | 1939 | |||
BC100819.2 | - | 16 | 1939 | |||
BC100820.1 | - | 16 | 1939 | |||
BC100821.2 | - | 16 | 1892 | |||
U78556.1 | - | 13 | 2748 | |||
U78557.1 | - | 9 | 2153 |