Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017369.2 | NP_001017369.1 | 5 | 738 | ||
NM_006745.4 | NP_006736.1 | 6 | 1024 | |||
XM_005263176.2 | XP_005263233.1 | 6 | 1141 | |||
GenBank mRNA | AK292418.1 | - | 6 | 969 | ||
AK295432.1 | - | 5 | 684 | |||
BC010653.1 | - | 5 | 918 | 3866053 | ||
BC107879.1 | - | 6 | 986 | 6498540 | ||
U60205.1 | - | 5 | 920 | |||
U93162.1 | - | 6 | 984 |