Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002912.4 | NP_001002912.4 | 15 | 6868 | ||
XM_017000274.1 | XP_016855763.1 | 14 | 7527 | |||
XM_017000276.1 | XP_016855765.1 | 4 | 5296 | |||
XM_017000277.1 | XP_016855766.1 | 3 | 5254 | |||
XM_017000278.1 | XP_016855767.1 | 4 | 5269 | |||
XM_017000279.1 | XP_016855768.1 | 4 | 5723 | |||
XM_017000280.1 | XP_016855769.1 | 3 | 5664 | |||
XM_017000281.1 | XP_016855770.1 | 2 | 6020 | |||
XM_017000282.1 | XP_016855771.1 | 3 | 5227 | |||
XM_017000283.1 | XP_016855772.1 | 3 | 5681 | |||
XM_017000284.1 | XP_016855773.1 | 2 | 5475 | |||
XM_017000285.1 | XP_016855774.1 | 2 | 5622 | |||
GenBank mRNA | AL833250.1 | - | 2 | 2176 | ||
AL834414.1 | - | 2 | 3935 | |||
BC035114.1 | - | 1 | 1701 | |||
BC044605.1 | - | 1 | 1928 | |||
CR749349.1 | - | 3 | 4792 |