Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DZIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014648.3 | NP_055463.1 | 6, 7 | 875 | ||
XM_005247914.3 | XP_005247971.1 | 6, 7 | 674 | |||
XM_005247915.3 | XP_005247972.1 | 6, 7 | 644 | |||
XM_005247916.3 | XP_005247973.1 | 7, 8 | 816 | |||
XM_005247917.3 | XP_005247974.1 | 6, 7 | 605 | |||
XM_005247918.3 | XP_005247975.1 | 6, 7 | 1092 | |||
XM_005247919.3 | XP_005247976.1 | 6, 7 | 844 | |||
XM_017007540.1 | XP_016863029.1 | 6, 7 | 721 | |||
GenBank mRNA | AB014575.1 | - | 6, 7 | 582 | ||
AK023138.1 | - | 6, 7 | 643 | |||
AK023389.1 | - | 6, 7 | 684 | |||
AY227651.1 | - | 6, 7 | 621 | |||
AY227654.1 | - | 6, 7 | 660 | |||
BC039018.1 | - | 6, 7 | 636 | |||
BC056674.1 | - | 6, 7 | 647 | |||
BC063882.1 | - | 6, 7 | 628 | 30387514 |